Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 611882 MIM: 605221 | |||||||||||||||||||||||||||||
Literature Links: |
PNRC2 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PNRC2 - proline rich nuclear receptor coactivator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRSF10 - serine and arginine rich splicing factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191005.2 | 2029 | UTR 3 | NP_001177934.1 | |||
NM_001191006.2 | 2029 | UTR 3 | NP_001177935.1 | |||
NM_001191007.2 | 2029 | UTR 3 | NP_001177936.1 | |||
NM_001191009.2 | 2029 | UTR 3 | NP_001177938.1 | |||
NM_001300936.1 | 2029 | UTR 3 | NP_001287865.1 | |||
NM_001300937.1 | 2029 | UTR 3 | NP_001287866.1 | |||
NM_006625.5 | 2029 | UTR 3 | NP_006616.1 | |||
NM_054016.3 | 2029 | UTR 3 | NP_473357.1 | |||
XM_017000100.1 | 2029 | Intron | XP_016855589.1 | |||
XM_017000101.1 | 2029 | Intron | XP_016855590.1 | |||
XM_017000102.1 | 2029 | Intron | XP_016855591.1 |
Set Membership: |
HapMap |