Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAATTGGTGACTGCAATAGCCA[A/T]GACCATGCTGAGCAAGGCTACACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR13G1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR13G1 - olfactory receptor family 13 subfamily G member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005487.1 | 436 | Missense Mutation | ATG,TTG | M,L 146 | NP_001005487.1 |