Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATCCAAACATCCAGCACTACTT[A/T]GTGTGGTCATTTTTGTGGTTTTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2T4 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
AMR
|
|||||
EUR - Not Available |
OR2T4 - olfactory receptor family 2 subfamily T member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004696.1 | Intron | NP_001004696.1 |