Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTAGTCCAAAGTGGTGTCCAT[C/T]CTTTAAGATTTTAAGATGGGGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBCE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBCE - tubulin folding cofactor E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079515.2 | Intron | NP_001072983.1 | ||||
NM_001287801.1 | Intron | NP_001274730.1 | ||||
NM_001287802.1 | Intron | NP_001274731.1 | ||||
NM_003193.4 | Intron | NP_003184.1 |