Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606435 MIM: 606429 MIM: 606430 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT1A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT1A10 - UDP glucuronosyltransferase family 1 member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019075.2 | 261 | Intron | NP_061948.1 |
UGT1A4 - UDP glucuronosyltransferase family 1 member A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UGT1A5 - UDP glucuronosyltransferase family 1 member A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019078.1 | 261 | Silent Mutation | GAC,GAT | D,D 87 | NP_061951.1 |
UGT1A6 - UDP glucuronosyltransferase family 1 member A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001072.3 | 261 | Intron | NP_001063.2 | |||
NM_205862.1 | 261 | Intron | NP_995584.1 |
UGT1A7 - UDP glucuronosyltransferase family 1 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019077.2 | 261 | Intron | NP_061950.2 |
UGT1A8 - UDP glucuronosyltransferase family 1 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019076.4 | 261 | Intron | NP_061949.3 |
UGT1A9 - UDP glucuronosyltransferase family 1 member A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021027.2 | 261 | Intron | NP_066307.1 |