Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCCAGGGCAGTTCCCTCAGCTC[C/T]GGGTTGGAGCCGGAGCCGGAGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607987 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC10 - DnaJ heat shock protein family (Hsp40) member C10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271581.1 | 51 | UTR 5 | NP_001258510.1 | |||
NM_018981.2 | 51 | UTR 5 | NP_061854.1 |