Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTTAGAAGTCTTGTCTTCAGAG[A/G]ACCTTACAGAATGATCCTGAGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604727 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK17B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STK17B - serine/threonine kinase 17b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004226.3 | 1283 | Missense Mutation | TCC,TTC | S,F 320 | NP_004217.1 | |
XM_011512168.2 | 1283 | Missense Mutation | TCC,TTC | S,F 320 | XP_011510470.1 | |
XM_011512169.1 | 1283 | Missense Mutation | TCC,TTC | S,F 320 | XP_011510471.1 | |
XM_011512170.1 | 1283 | Missense Mutation | TCC,TTC | S,F 320 | XP_011510472.1 | |
XM_011512171.2 | 1283 | Missense Mutation | TCC,TTC | S,F 192 | XP_011510473.1 |
Set Membership: |
HapMap |