Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGTTTTATCCAACTTTAGCCC[A/C]AGGTGGTACACTGGCAGATGACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BZW1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BZW1 - basic leucine zipper and W2 domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207067.1 | 700 | Missense Mutation | CAA,CCA | Q,P 82 | NP_001193996.1 | |
NM_001207069.1 | 700 | Intron | NP_001193998.1 | |||
NM_001321688.1 | 700 | Intron | NP_001308617.1 | |||
NM_001321690.1 | 700 | Intron | NP_001308619.1 | |||
NM_001321691.1 | 700 | Intron | NP_001308620.1 | |||
NM_001321693.1 | 700 | Intron | NP_001308622.1 | |||
NM_001321694.1 | 700 | Intron | NP_001308623.1 | |||
NM_014670.3 | 700 | Missense Mutation | CAA,CCA | Q,P 82 | NP_055485.2 | |
XM_017005393.1 | 700 | Intron | XP_016860882.1 |
LOC101927795 - uncharacterized LOC101927795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |