Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTCAAATCAGTTTCTTTCTAAC[A/G]AAATACGTCAAGGTAGAAGAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CR12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALS2CR12 - amyotrophic lateral sclerosis 2 chromosome region candidate 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127391.2 | 1460 | UTR 3 | NP_001120863.1 | |||
NM_001289993.1 | 1460 | UTR 3 | NP_001276922.1 | |||
NM_139163.3 | 1460 | UTR 3 | NP_631902.2 | |||
XM_011510606.2 | 1460 | UTR 3 | XP_011508908.1 | |||
XM_011510610.2 | 1460 | UTR 3 | XP_011508912.1 | |||
XM_011510612.2 | 1460 | UTR 3 | XP_011508914.1 | |||
XM_017003360.1 | 1460 | Intron | XP_016858849.1 | |||
XM_017003361.1 | 1460 | Intron | XP_016858850.1 | |||
XM_017003362.1 | 1460 | UTR 3 | XP_016858851.1 |
CASP8 - caspase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080124.1 | 1460 | Intron | NP_001073593.1 | |||
NM_001080125.1 | 1460 | Intron | NP_001073594.1 | |||
NM_001228.4 | 1460 | Intron | NP_001219.2 | |||
NM_033355.3 | 1460 | Intron | NP_203519.1 | |||
NM_033356.3 | 1460 | Intron | NP_203520.1 | |||
NM_033358.3 | 1460 | Intron | NP_203522.1 | |||
XM_005246885.1 | 1460 | Intron | XP_005246942.1 | |||
XM_005246886.1 | 1460 | Intron | XP_005246943.1 | |||
XM_005246887.1 | 1460 | Intron | XP_005246944.1 | |||
XM_005246888.1 | 1460 | Intron | XP_005246945.1 | |||
XM_005246889.1 | 1460 | Intron | XP_005246946.1 | |||
XM_005246890.3 | 1460 | Intron | XP_005246947.1 | |||
XM_005246891.4 | 1460 | Intron | XP_005246948.1 | |||
XM_005246892.1 | 1460 | Intron | XP_005246949.1 | |||
XM_005246893.2 | 1460 | Intron | XP_005246950.1 | |||
XM_005246894.3 | 1460 | Intron | XP_005246951.1 | |||
XM_005246895.2 | 1460 | Intron | XP_005246952.1 | |||
XM_006712789.1 | 1460 | Intron | XP_006712852.1 | |||
XM_006712790.3 | 1460 | Intron | XP_006712853.1 | |||
XM_006712793.2 | 1460 | Intron | XP_006712856.1 | |||
XM_011511969.1 | 1460 | Intron | XP_011510271.1 |