Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTAGAGTGACATACCTCCCTCCC[A/G]AAAACACCTGTGCCTTGACATAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALS2CR12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALS2CR12 - amyotrophic lateral sclerosis 2 chromosome region candidate 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127391.2 | Intron | NP_001120863.1 | ||||
NM_001289993.1 | Intron | NP_001276922.1 | ||||
NM_139163.3 | Intron | NP_631902.2 | ||||
XM_011510606.2 | Intron | XP_011508908.1 | ||||
XM_011510610.2 | Intron | XP_011508912.1 | ||||
XM_011510612.2 | Intron | XP_011508914.1 | ||||
XM_017003360.1 | Intron | XP_016858849.1 | ||||
XM_017003361.1 | Intron | XP_016858850.1 | ||||
XM_017003362.1 | Intron | XP_016858851.1 |
CASP8 - caspase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |