Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATTCGGGGCTGTTTGGGTCTA[A/G]CTTGGTCTAGACGGGTCTGAAACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146731 | ||||||||||||||||||||
Literature Links: |
IGFBP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGFBP2 - insulin like growth factor binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000597.2 | 55 | Intron | NP_000588.2 | |||
NM_001313990.1 | 55 | Intron | NP_001300919.1 | |||
NM_001313992.1 | 55 | UTR 5 | NP_001300921.1 | |||
NM_001313993.1 | 55 | UTR 5 | NP_001300922.1 |