Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610405 MIM: 610991 | ||||||||||||||||||||
Literature Links: |
CHPF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHPF - chondroitin polymerizing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3132 - microRNA 3132 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OBSL1 - obscurin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173408.1 | 5994 | Intron | NP_001166879.1 | |||
NM_001173431.1 | 5994 | Intron | NP_001166902.1 | |||
NM_015311.2 | 5994 | Missense Mutation | AAG,GAG | K,E 1894 | NP_056126.1 | |
XM_005246424.4 | 5994 | Missense Mutation | AAG,GAG | K,E 1802 | XP_005246481.1 | |
XM_005246427.4 | 5994 | Intron | XP_005246484.1 | |||
XM_011510857.2 | 5994 | Missense Mutation | AAG,GAG | K,E 1894 | XP_011509159.1 | |
XM_011510863.2 | 5994 | Intron | XP_011509165.1 | |||
XM_011510864.2 | 5994 | Intron | XP_011509166.1 | |||
XM_011510865.2 | 5994 | Intron | XP_011509167.1 | |||
XM_011510866.2 | 5994 | Intron | XP_011509168.1 | |||
XM_017003696.1 | 5994 | Missense Mutation | AAG,GAG | K,E 1894 | XP_016859185.1 | |
XM_017003697.1 | 5994 | Missense Mutation | AAG,GAG | K,E 1894 | XP_016859186.1 | |
XM_017003698.1 | 5994 | Missense Mutation | AAG,GAG | K,E 1802 | XP_016859187.1 | |
XM_017003699.1 | 5994 | Missense Mutation | AAG,GAG | K,E 1802 | XP_016859188.1 | |
XM_017003700.1 | 5994 | Missense Mutation | AAG,GAG | K,E 1710 | XP_016859189.1 |
TMEM198 - transmembrane protein 198 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005209.2 | 5994 | Intron | NP_001005209.1 | |||
NM_001303098.1 | 5994 | Intron | NP_001290027.1 | |||
XM_017003367.1 | 5994 | Intron | XP_016858856.1 |