Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTATTAATTAAATAAAGGCTCCC[A/G]GCTTTTGGTGGAAAACATTGCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A14 - solute carrier family 16 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152527.4 | Intron | NP_689740.2 | ||||
XM_005246353.3 | Intron | XP_005246410.1 | ||||
XM_011510751.2 | Intron | XP_011509053.1 | ||||
XM_011510752.1 | Intron | XP_011509054.1 | ||||
XM_017003480.1 | Intron | XP_016858969.1 |