Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188390 | ||||||||||||||||||||
Literature Links: |
MIR1244-1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR1244-1 - microRNA 1244-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTMA - prothymosin, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099285.1 | 973 | Intron | NP_001092755.1 | |||
NM_002823.4 | 973 | Intron | NP_002814.3 | |||
XM_011511518.1 | 973 | Missense Mutation | GAA,GAC | E,D 207 | XP_011509820.1 | |
XM_011511519.1 | 973 | Missense Mutation | GAA,GAC | E,D 206 | XP_011509821.1 | |
XM_011511520.1 | 973 | Missense Mutation | GAA,GAC | E,D 206 | XP_011509822.1 | |
XM_011511521.1 | 973 | Missense Mutation | GAA,GAC | E,D 205 | XP_011509823.1 | |
XM_011511522.2 | 973 | Intron | XP_011509824.1 |