Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGGGTCTAATCAGACCCAAAAA[C/T]ACGAAGCCCAGAAACCCCCAAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602255 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FARP2 - FERM, ARH/RhoGEF and pleckstrin domain protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK25 - serine/threonine kinase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271977.1 | 1845 | Intron | NP_001258906.1 | |||
NM_001271978.1 | 1845 | Intron | NP_001258907.1 | |||
NM_001271979.1 | 1845 | Intron | NP_001258908.1 | |||
NM_001271980.1 | 1845 | Intron | NP_001258909.1 | |||
NM_001282305.1 | 1845 | Intron | NP_001269234.1 | |||
NM_001282306.1 | 1845 | Intron | NP_001269235.1 | |||
NM_001282307.1 | 1845 | Intron | NP_001269236.1 | |||
NM_001282308.1 | 1845 | Intron | NP_001269237.1 | |||
NM_006374.4 | 1845 | Intron | NP_006365.2 | |||
XM_011510493.2 | 1845 | UTR 3 | XP_011508795.1 | |||
XM_011510494.2 | 1845 | Intron | XP_011508796.1 | |||
XM_011510495.2 | 1845 | Intron | XP_011508797.1 | |||
XM_011510496.2 | 1845 | UTR 3 | XP_011508798.1 | |||
XM_017003168.1 | 1845 | Intron | XP_016858657.1 | |||
XM_017003169.1 | 1845 | Intron | XP_016858658.1 | |||
XM_017003170.1 | 1845 | Intron | XP_016858659.1 | |||
XM_017003171.1 | 1845 | Intron | XP_016858660.1 |