Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAATCTAATTGTTGAAACAGGGA[A/T]AAACAGCTCATGCTTGAATCATTGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607659 MIM: 602339 | |||||||||||||||||||||||
Literature Links: |
EAF2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EAF2 - ELL associated factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC15A2 - solute carrier family 15 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145998.1 | Intron | NP_001139470.1 | ||||
NM_021082.3 | Intron | NP_066568.3 | ||||
XM_005247722.3 | Intron | XP_005247779.1 | ||||
XM_006713736.3 | Intron | XP_006713799.1 | ||||
XM_017007074.1 | Intron | XP_016862563.1 |