Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGACCTGTTCGCCAGCTGCAGGAA[A/G]GGGGATGTGGGCCGAGTGCGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608308 MIM: 616627 | ||||||||||||||||||||
Literature Links: |
ABTB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABTB1 - ankyrin repeat and BTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032548.3 | 121 | UTR 5 | NP_115937.1 | |||
NM_172027.2 | 121 | Missense Mutation | AAG,AAT | K,N 12 | NP_742024.1 | |
XM_006713769.3 | 121 | UTR 5 | XP_006713832.1 | |||
XM_011513213.1 | 121 | Intron | XP_011511515.1 | |||
XM_017007285.1 | 121 | Silent Mutation | AAA,AAG | K,K 12 | XP_016862774.1 | |
XM_017007286.1 | 121 | UTR 5 | XP_016862775.1 | |||
XM_017007287.1 | 121 | Intron | XP_016862776.1 | |||
XM_017007288.1 | 121 | UTR 5 | XP_016862777.1 |
PODXL2 - podocalyxin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015720.3 | 121 | Intron | NP_056535.1 |