Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAAAATACCTGAGAAGGGTTCT[A/T]CAAAATTTCAATATCATGAAATGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603273 | ||||||||||||||||||||
Literature Links: |
TP63 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TP63 - tumor protein p63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114978.1 | Intron | NP_001108450.1 | ||||
NM_001114979.1 | Intron | NP_001108451.1 | ||||
NM_001114980.1 | Intron | NP_001108452.1 | ||||
NM_001114981.1 | Intron | NP_001108453.1 | ||||
NM_001114982.1 | Intron | NP_001108454.1 | ||||
NM_003722.4 | Intron | NP_003713.3 | ||||
XM_005247843.3 | Intron | XP_005247900.1 | ||||
XM_005247844.4 | Intron | XP_005247901.1 | ||||
XM_011513251.2 | Intron | XP_011511553.1 | ||||
XM_011513252.2 | Intron | XP_011511554.1 | ||||
XM_017007387.1 | Intron | XP_016862876.1 |