Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACCTCAGCGTGTCCCCACTGCC[A/G]CCACACCTCCGACATACCTATTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602462 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf50 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf50 - chromosome 4 open reading frame 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011513603.2 | 11032 | UTR 3 | XP_011511905.1 | |||
XM_011513604.2 | 11032 | UTR 3 | XP_011511906.1 | |||
XM_017008891.1 | 11032 | UTR 3 | XP_016864380.1 | |||
XM_017008892.1 | 11032 | UTR 3 | XP_016864381.1 | |||
XM_017008893.1 | 11032 | UTR 3 | XP_016864382.1 | |||
XM_017008894.1 | 11032 | Intron | XP_016864383.1 |
CRMP1 - collapsin response mediator protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |