Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAGAAGGGACCCACTAGGCACGG[A/G]AGAGGCCGGCCCGTGCGGGCAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFR3 - fibroblast growth factor receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000142.4 | Intron | NP_000133.1 | ||||
NM_001163213.1 | Intron | NP_001156685.1 | ||||
NM_022965.3 | Intron | NP_075254.1 | ||||
XM_006713868.1 | Intron | XP_006713931.1 | ||||
XM_006713869.1 | Intron | XP_006713932.1 | ||||
XM_006713870.1 | Intron | XP_006713933.1 | ||||
XM_006713871.1 | Intron | XP_006713934.1 | ||||
XM_006713872.1 | Intron | XP_006713935.1 | ||||
XM_006713873.1 | Intron | XP_006713936.1 | ||||
XM_011513420.1 | Intron | XP_011511722.1 | ||||
XM_011513422.1 | Intron | XP_011511724.1 |