Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGCTCCTGCTTGCGAAACTTGGC[C/G]CGGCGGTTCTGGAACCACACCTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHOX2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHOX2B - paired like homeobox 2b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003924.3 | 810 | Silent Mutation | CGC,CGG | R,R 150 | NP_003915.2 |