Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTTCTGAAGCTCAGCGACATC[C/T]CCTGGCAAGACTCTGAACTGGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612994 | ||||||||||||||||||||
Literature Links: |
RAB28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAB28 - RAB28, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017979.2 | Intron | NP_001017979.1 | ||||
NM_001159601.1 | Intron | NP_001153073.1 | ||||
NM_004249.3 | Intron | NP_004240.2 | ||||
XM_005248218.4 | Intron | XP_005248275.1 | ||||
XM_011513911.2 | Intron | XP_011512213.1 | ||||
XM_011513912.2 | Intron | XP_011512214.1 | ||||
XM_017008850.1 | Intron | XP_016864339.1 |