Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGGGCAGTCAGCGTCGTCCC[C/T]GGGAGCTTCCGACTCAGCTGCGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610300 MIM: 187270 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A18 - solute carrier family 6 member 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TERT - telomerase reverse transcriptase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193376.1 | 3193 | Silent Mutation | CCA,CCG | P,P 1045 | NP_001180305.1 | |
NM_198253.2 | 3193 | Silent Mutation | CCA,CCG | P,P 1108 | NP_937983.2 | |
XM_017009796.1 | 3193 | Silent Mutation | CCA,CCG | P,P 1318 | XP_016865285.1 |