Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTATTAAGACAGTTTAATATTA[A/G]TGAAATTCTTTAGCCATTTCATTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600946 | ||||||||||||||||||||
Literature Links: |
GHR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GHR - growth hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000163.4 | Intron | NP_000154.1 | ||||
NM_001242399.2 | Intron | NP_001229328.1 | ||||
NM_001242400.2 | Intron | NP_001229329.1 | ||||
NM_001242401.3 | Intron | NP_001229330.1 | ||||
NM_001242402.2 | Intron | NP_001229331.1 | ||||
NM_001242403.2 | Intron | NP_001229332.1 | ||||
NM_001242404.2 | Intron | NP_001229333.1 | ||||
NM_001242405.2 | Intron | NP_001229334.1 | ||||
NM_001242406.2 | Intron | NP_001229335.1 | ||||
NM_001242460.1 | Intron | NP_001229389.1 | ||||
NM_001242462.1 | Intron | NP_001229391.1 |