Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTACACTGTCAGGTGATTGCAG[A/G]CCCTGTTTTTTCAAATATCAGATGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601484 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC152 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC152 - coiled-coil domain containing 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134848.1 | 1980 | UTR 3 | NP_001128320.1 |
SEPP1 - selenoprotein P, plasma, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085486.1 | 1980 | Missense Mutation | CCT,TCT | P,S 314 | NP_001078955.1 | |
NM_001093726.1 | 1980 | Missense Mutation | CCT,TCT | P,S 344 | NP_001087195.1 | |
NM_005410.2 | 1980 | Missense Mutation | CCT,TCT | P,S 314 | NP_005401.3 |