Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTCCTCTATCACACCTTCACC[A/G]GGTGCTGTGGGAAAGTGCCTGTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRD9 - bromodomain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009877.2 | Intron | NP_001009877.2 | ||||
NM_001317951.1 | Intron | NP_001304880.1 | ||||
NM_023924.4 | Intron | NP_076413.3 | ||||
XM_006714494.3 | Intron | XP_006714557.1 | ||||
XM_017009780.1 | Intron | XP_016865269.1 |
ZDHHC11 - zinc finger DHHC-type containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |