Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTTGTCACGGCACCGCCTACCA[A/G]GACGGGCGGTTAAGACACTAGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194363 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM167A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM167A - transmembrane protein 167A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174909.4 | 42 | Intron | NP_777569.1 |
XRCC4 - X-ray repair cross complementing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318012.1 | 42 | UTR 5 | NP_001304941.1 | |||
NM_001318013.1 | 42 | UTR 5 | NP_001304942.1 | |||
NM_003401.4 | 42 | UTR 5 | NP_003392.1 | |||
NM_022406.3 | 42 | UTR 5 | NP_071801.1 | |||
NM_022550.3 | 42 | UTR 5 | NP_072044.1 | |||
XM_011543626.1 | 42 | Intron | XP_011541928.1 | |||
XM_017009827.1 | 42 | UTR 5 | XP_016865316.1 | |||
XM_017009828.1 | 42 | UTR 5 | XP_016865317.1 | |||
XM_017009829.1 | 42 | UTR 5 | XP_016865318.1 |