Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGCCGCCGCCCCACGGAGCCGC[C/G]AGCCGCGGCGGCGCCCACCTTACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600982 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K1 - mitogen-activated protein kinase kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005921.1 | 852 | Silent Mutation | GCC,GCG | A,A 117 | NP_005912.1 | |
XM_017009484.1 | 852 | Intron | XP_016864973.1 | |||
XM_017009485.1 | 852 | Intron | XP_016864974.1 |