Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTAGTTGTTTTTTGCATTGCTG[A/G]TAGTAAGTAGAAAGGCTTTTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602614 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K7 - mitogen-activated protein kinase kinase kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003188.3 | 1961 | Silent Mutation | TAC,TAT | Y,Y 558 | NP_003179.1 | |
NM_145331.2 | 1961 | Silent Mutation | TAC,TAT | Y,Y 585 | NP_663304.1 | |
NM_145332.2 | 1961 | UTR 3 | NP_663305.1 | |||
NM_145333.2 | 1961 | UTR 3 | NP_663306.1 | |||
XM_006715553.2 | 1961 | Silent Mutation | TAC,TAT | Y,Y 455 | XP_006715616.1 | |
XM_017011226.1 | 1961 | Silent Mutation | TAC,TAT | Y,Y 428 | XP_016866715.1 |