Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGAGTCCAGGAAGGGGGTGGAG[C/T]GTGGCCATGGATAATCGGGCTTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115441 MIM: 142610 MIM: 610433 MIM: 610434 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK2B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CSNK2B - casein kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPANK1 - G-patch domain and ankyrin repeats 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6G5B - lymphocyte antigen 6 complex, locus G5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021221.2 | Intron | NP_067044.2 |
LY6G5C - lymphocyte antigen 6 complex, locus G5C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |