Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTGGTGCTGCAGGGTCTGGGTC[C/T]TTGCAGGATACACAGTCACCTTAGG
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604776 | |||||||||||||||||||||||||||||
Literature Links: |
HLA-DRB5 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
SAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
EAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
EUR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
AFR - Not Available | |||||
HLA-DRB5 - major histocompatibility complex, class II, DR beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002125.3 | 406 | Missense Mutation | AAG,AGG | K,R 134 | NP_002116.2 | |
XM_011514562.2 | 406 | Intron | XP_011512864.1 |