Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGGACTACTAGGTGCTGATTCC[G/T]GGAATTTCCTGCAATGTCCCTGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604100 | ||||||||||||||||||||
Literature Links: |
GRM4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRM4 - glutamate metabotropic receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000841.3 | 4427 | UTR 3 | NP_000832.1 | |||
NM_001256809.2 | 4427 | UTR 3 | NP_001243738.1 | |||
NM_001256811.2 | 4427 | UTR 3 | NP_001243740.1 | |||
NM_001256812.2 | 4427 | UTR 3 | NP_001243741.1 | |||
NM_001256813.2 | 4427 | UTR 3 | NP_001243742.1 | |||
NM_001282847.1 | 4427 | UTR 3 | NP_001269776.1 | |||
XM_017010790.1 | 4427 | UTR 3 | XP_016866279.1 | |||
XM_017010791.1 | 4427 | UTR 3 | XP_016866280.1 | |||
XM_017010792.1 | 4427 | UTR 3 | XP_016866281.1 | |||
XM_017010793.1 | 4427 | UTR 3 | XP_016866282.1 |