Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCCCCATGCTCTTCATGCCAGC[A/G]TAGGGGTGACCTAGCAGGGCCCTCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140572 MIM: 604548 MIM: 610788 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSP90AB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HSP90AB1 - heat shock protein 90 alpha family class B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4647 - microRNA 4647 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIE - NFKB inhibitor epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004556.2 | 2459 | UTR 3 | NP_004547.2 |
SLC35B2 - solute carrier family 35 member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286509.1 | 2459 | Intron | NP_001273438.1 | |||
NM_001286510.1 | 2459 | Intron | NP_001273439.1 | |||
NM_001286511.1 | 2459 | Intron | NP_001273440.1 | |||
NM_001286512.1 | 2459 | Intron | NP_001273441.1 | |||
NM_001286513.1 | 2459 | Intron | NP_001273442.1 | |||
NM_001286517.1 | 2459 | Intron | NP_001273446.1 | |||
NM_001286519.1 | 2459 | Intron | NP_001273448.1 | |||
NM_001286520.1 | 2459 | Intron | NP_001273449.1 | |||
NM_178148.3 | 2459 | Intron | NP_835361.1 |
Set Membership: |
HapMap |