Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAACTTAATGTTGGTAGAGCTG[A/G]GAATAGCAGTTTCCTGGTAGAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604756 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC441239 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC441239 - uncharacterized LOC441239 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF273 - zinc finger protein 273 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021148.2 | Intron | NP_066971.2 | ||||
XM_011515746.2 | Intron | XP_011514048.1 | ||||
XM_017011688.1 | Intron | XP_016867177.1 | ||||
XM_017011689.1 | Intron | XP_016867178.1 | ||||
XM_017011690.1 | Intron | XP_016867179.1 | ||||
XM_017011691.1 | Intron | XP_016867180.1 | ||||
XM_017011692.1 | Intron | XP_016867181.1 | ||||
XM_017011693.1 | Intron | XP_016867182.1 | ||||
XM_017011694.1 | Intron | XP_016867183.1 | ||||
XM_017011695.1 | Intron | XP_016867184.1 | ||||
XM_017011696.1 | Intron | XP_016867185.1 | ||||
XM_017011697.1 | Intron | XP_016867186.1 | ||||
XM_017011698.1 | Intron | XP_016867187.1 |