Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGGCTCGGACGCCAGCGGACAC[C/G]CGGGCGAGGTACGGAGTGGACGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609700 | ||||||||||||||||||||
Literature Links: |
RABGEF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RABGEF1 - RAB guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287060.1 | 160 | UTR 5 | NP_001273989.1 | |||
NM_001287061.1 | 160 | Intron | NP_001273990.1 | |||
NM_001287062.1 | 160 | Intron | NP_001273991.1 | |||
NM_014504.2 | 160 | Intron | NP_055319.1 |