Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGAGACAAGGAGAGCGGGGCGG[A/G]GGCTTGGGGGAGACCAGGAGTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602686 MIM: 606906 MIM: 600312 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAD1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAD1L1 - MAD1 mitotic arrest deficient like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRM2 - mitochondrial rRNA methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013393.1 | Intron | NP_037525.1 |
NUDT1 - nudix hydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002452.3 | Intron | NP_002443.3 | ||||
NM_198948.1 | Intron | NP_945186.1 | ||||
NM_198949.1 | Intron | NP_945187.1 | ||||
NM_198950.1 | Intron | NP_945188.1 | ||||
NM_198952.1 | Intron | NP_945190.1 | ||||
NM_198953.1 | Intron | NP_945191.1 | ||||
NM_198954.1 | Intron | NP_945192.1 |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |