Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCTGTTTCTACCCCAGAAAATC[A/C]CTGGTTCAATGGCAGTTTTCCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606906 MIM: 600312 MIM: 614905 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRM2 - mitochondrial rRNA methyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT1 - nudix hydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002452.3 | Intron | NP_002443.3 | ||||
NM_198948.1 | Intron | NP_945186.1 | ||||
NM_198949.1 | Intron | NP_945187.1 | ||||
NM_198950.1 | Intron | NP_945188.1 | ||||
NM_198952.1 | Intron | NP_945190.1 | ||||
NM_198953.1 | Intron | NP_945191.1 | ||||
NM_198954.1 | Intron | NP_945192.1 |
SNX8 - sorting nexin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |