Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTATGGTGAGTGTGTCCCCAGG[A/G]CTGGTAGTCACCTGGCTGTCCGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD11 - caspase recruitment domain family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324281.1 | 4259 | UTR 3 | NP_001311210.1 | |||
NM_032415.5 | 4259 | UTR 3 | NP_115791.3 | |||
XM_011515586.2 | 4259 | UTR 3 | XP_011513888.1 | |||
XM_011515587.2 | 4259 | UTR 3 | XP_011513889.1 |