Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTAGAAGATTATATGGAGGAGG[A/G]AACGGGTGTGGAGGTGCTCGGGTGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 616068 MIM: 142954 MIM: 142953 | |||||||||||||||||||||||
Literature Links: |
HOXA-AS2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HOXA-AS2 - HOXA cluster antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA3 - homeobox A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030661.4 | 996 | Intron | NP_109377.1 | |||
NM_153631.2 | 996 | Intron | NP_705895.1 | |||
XM_005249730.2 | 996 | Intron | XP_005249787.1 | |||
XM_005249731.2 | 996 | Intron | XP_005249788.1 | |||
XM_005249732.3 | 996 | Intron | XP_005249789.1 | |||
XM_006715715.2 | 996 | Intron | XP_006715778.1 | |||
XM_011515343.2 | 996 | Intron | XP_011513645.1 |
HOXA4 - homeobox A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002141.4 | 996 | Missense Mutation | CCC,TCC | P,S 317 | NP_002132.3 |