Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTACCCAATCAAGGGCAGGCCC[C/T]TATTTTCCCTTCTTGGGTTCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142996 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EVX1 - even-skipped homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304519.1 | Intron | NP_001291448.1 | ||||
NM_001304520.1 | Intron | NP_001291449.1 | ||||
NM_001989.4 | Intron | NP_001980.1 |
EVX1-AS - EVX1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |