Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTAGCCAGTTTAGACGTGGCCG[A/C]GGGGGATGTACGGGACAGAGCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP1 - aquaporin 1 (Colton blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185060.1 | Intron | NP_001171989.1 | ||||
NM_001185061.1 | Intron | NP_001171990.1 | ||||
NM_001185062.1 | Intron | NP_001171991.1 |