Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGTGCAGCCCCCATACCGGGACG[A/T]GGCCTCGCCCGTCCCACCCCGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607723 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUN1 - Sad1 and UNC84 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130965.2 | Intron | NP_001124437.1 | ||||
NM_001171944.1 | Intron | NP_001165415.1 | ||||
NM_001171945.1 | Intron | NP_001165416.1 | ||||
NM_001171946.1 | Intron | NP_001165417.1 | ||||
NM_025154.5 | Intron | NP_079430.3 |