Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATCCATATGAGACAGAATTCTCT[C/T]GGTAATAACAGGCCCTTCAGATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR2J4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR2J4 - RNA polymerase II subunit J4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2D4 - ubiquitin conjugating enzyme E2 D4 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015983.3 | Intron | NP_057067.1 | ||||
XM_006715734.3 | Intron | XP_006715797.1 | ||||
XM_011515420.2 | Intron | XP_011513722.1 | ||||
XM_011515422.2 | Intron | XP_011513724.1 | ||||
XM_011515423.2 | Intron | XP_011513725.1 | ||||
XM_017012292.1 | Intron | XP_016867781.1 | ||||
XM_017012293.1 | Intron | XP_016867782.1 | ||||
XM_017012294.1 | Intron | XP_016867783.1 | ||||
XM_017012295.1 | Intron | XP_016867784.1 |