Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAAGATGAAGAATTAACTATAA[C/T]ATATAAGAGACTTATTAACACTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPA3 - replication protein A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002947.4 | Intron | NP_002938.1 |
UMAD1 - UBAP1-MVB12-associated (UMA) domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |