Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGCCTTTTTAAGCCTGAATTT[C/T]CATCGTCTTAGAGATGAGTGAGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF862 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF862 - zinc finger protein 862 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099220.1 | Intron | NP_001092690.1 | ||||
XM_006716085.3 | Intron | XP_006716148.1 | ||||
XM_011516463.1 | Intron | XP_011514765.1 | ||||
XM_011516464.1 | Intron | XP_011514766.1 | ||||
XM_011516465.2 | Intron | XP_011514767.1 | ||||
XM_011516466.2 | Intron | XP_011514768.1 | ||||
XM_017012519.1 | Intron | XP_016868008.1 |