Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCGTGTCAATGTTATACACACGT[A/C]AGCACACAGAGCAGCCAGGATGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ACTR3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR3C - ARP3 actin-related protein 3 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164458.1 | Intron | NP_001157930.1 | ||||
NM_001164459.1 | Intron | NP_001157931.1 | ||||
XM_005250043.4 | Intron | XP_005250100.3 | ||||
XM_011516502.2 | Intron | XP_011514804.1 | ||||
XM_011516506.2 | Intron | XP_011514808.1 | ||||
XM_011516507.2 | Intron | XP_011514809.1 | ||||
XM_011516508.2 | Intron | XP_011514810.1 | ||||
XM_011516511.2 | Intron | XP_011514813.1 | ||||
XM_017012548.1 | Intron | XP_016868037.1 | ||||
XM_017012549.1 | Intron | XP_016868038.1 | ||||
XM_017012550.1 | Intron | XP_016868039.1 | ||||
XM_017012551.1 | Intron | XP_016868040.1 | ||||
XM_017012552.1 | Intron | XP_016868041.1 | ||||
XM_017012553.1 | Intron | XP_016868042.1 |