Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGCCCCGTTTGTTGTCTCGGAG[C/G]CCACCCGACTCTAGCCGCTGAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611741 MIM: 123831 MIM: 109280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC3 - acid sensing ion channel subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDK5 - cyclin dependent kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164410.2 | 376 | Intron | NP_001157882.1 | |||
NM_004935.3 | 376 | Intron | NP_004926.1 |
SLC4A2 - solute carrier family 4 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199692.1 | 376 | UTR 5 | NP_001186621.1 | |||
NM_001199693.1 | 376 | Intron | NP_001186622.1 | |||
NM_001199694.1 | 376 | Intron | NP_001186623.1 | |||
NM_003040.3 | 376 | Intron | NP_003031.3 | |||
XM_006716094.3 | 376 | Intron | XP_006716157.1 |