Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTCTGCCTCCAAAATTATCCTT[C/T]GAAAAACATCCACAGCAGTCTGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601293 | ||||||||||||||||||||
Literature Links: |
RHEB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHEB - Ras homolog enriched in brain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005614.3 | 949 | Missense Mutation | CAA,CGA | Q,R 161 | NP_005605.1 | |
XM_011516457.1 | 949 | Missense Mutation | CAA,CGA | Q,R 150 | XP_011514759.1 |