Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAGTCCTCCTAAGCTTAATTT[T/C]AGGGGGATCTGCAACAGTTAAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 124010 | ||||||||||||||||||||
Literature Links: |
CYP3A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP3A4 - cytochrome P450 family 3 subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202855.2 | 1529 | Silent Mutation | CTA,CTG | L,L 474 | NP_001189784.1 | |
NM_017460.5 | 1529 | Silent Mutation | CTA,CTG | L,L 475 | NP_059488.2 | |
XM_017011780.1 | 1529 | Intron | XP_016867269.1 |