Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCCAGCACCTTCTATGGTTCCA[G/T]TTGTGAGTTTGCTGTGGAACAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604609 MIM: 158371 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUC12 - mucin 12, cell surface associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MUC3A - mucin 3A, cell surface associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005960.1 | 8843 | Missense Mutation | AGT,ATT | S,I 3007 | NP_005951.1 | |
XM_011516246.2 | 8843 | Missense Mutation | AGT,ATT | S,I 2997 | XP_011514548.1 | |
XM_011516247.2 | 8843 | Missense Mutation | AGT,ATT | S,I 2980 | XP_011514549.1 | |
XM_011516249.2 | 8843 | Missense Mutation | AGT,ATT | S,I 2929 | XP_011514551.1 | |
XM_011516250.2 | 8843 | Missense Mutation | AGT,ATT | S,I 2760 | XP_011514552.1 | |
XM_017012231.1 | 8843 | Missense Mutation | AGT,ATT | S,I 2827 | XP_016867720.1 |